EMAIL THIS PAGE TO A FRIEND
Cell and tissue research
Localization of reelin signaling pathway components in murine midbrain and striatum.
PMID 25418135
Abstract
We investigated the distribution patterns of the extracellular matrix protein Reelin and of crucial Reelin signaling components in murine midbrain and striatum. The cellular distribution of the Reelin receptors VLDLr and ApoER2, the intracellular downstream mediator Dab1, and the alternative Reelin receptor APP were analyzed at embryonic day 16, at postnatal stage 15 (P15), and in 3-month-old mice. Reelin was expressed intracellularly and extracellularly in midbrain mesencephalic dopaminergic (mDA) neurons of newborns. In the striatum, Calbindin D-28k(+) neurons exhibited Reelin intracellularly at E16 and extracellularly at P15 and 3 months. ApoER2 and VLDLr were expressed in mDA neurons at E16 and P15 and in oligodendrocytes at 3 months, whereas Dab1 and APP immunoreactivity was observed in mDA at all stages analyzed. In the striatum, Calbindin D-28k(+)/GAD67(+) inhibitory neurons expressed VLDLr, ApoER2, and Dab1 at P15, but only Dab1 at E16 and 3 months. APP was always expressed in mouse striatum in which it colocalized with Calbindin D-28k. Our data underline the importance of Reelin signalling during embryonic development and early postnatal maturation of the mesostriatal and mesocorticolimbic system, and suggest that the striatum and not the midbrain is the primary source of Reelin for midbrain neurons. The loss of ApoER2 and VLDLr expression in the mature midbrain and striatum implies that Reelin functions are restricted to migratory events and early postnatal maturation and are dispensable for the maintenance of dopaminergic neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB348
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, 50 µg, Anti-APP A4 Antibody, a.a. 66-81 of APP {N-terminus}, clone 22C11 is an antibody against Alzheimer Precursor Protein A4 for use in IF, IH, IH(P) & WB with more than 85 product citations.
AB1778
Anti-Calbindin D-28K Antibody, 100 µL, This Anti-Calbindin D-28K Antibody is validated for use in IC, IH, IH(P), WB for the detection of Calbindin D-28K.
MAB5406
Anti-GAD67 Antibody, clone 1G10.2, 100 µg, This Anti-GAD Antibody, clone 1G10.2 is validated for use in IH, IH(P), WB for the detection of GAD67. This Anti GAD antibody has more than 75 product citations.
MAB3402
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, 40 µg, Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5 is an antibody against Glial Fibrillary Acidic Protein for use in IC, IH, IH(P) & WB with more than 55 product citations.
MAB377
Anti-NeuN Antibody, clone A60, 500 µg, Anti-NeuN Antibody, clone A60 detects level of NeuN and has been published and validated for use in FC, IC, IF, IH, IH(P), IP and WB.
AB9610
Anti-Olig-2 Antibody, 100 µL, Anti-Olig-2 Antibody is an antibody against Oligodendrocyte transcription factor 2 for use in IC, IH, IH(P), IP and WB.
MAB5364
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, 100 µg, Detect Reelin using this Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10 validated for use in IH & WB.
AB152
Anti-Tyrosine Hydroxylase Antibody, 100 µL, Anti-Tyrosine Hydroxylase Antibody detects level of TH and has been published and validated for use in ELISA, IF, IH, IH(P), IP and WB.
MAB318
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, 100 µL, Detect Tyrosine Hydroxylase using this Anti-Tyrosine Hydroxylase Antibody, clone LNC1 validated for use in IH, IH(P), IP & WB with more than 85 product citations.