EMAIL THIS PAGE TO A FRIEND
The AAPS journal
Safety and biocompatibility of carbohydrate-functionalized polyanhydride nanoparticles.
PMID 25421457
Abstract
Carbohydrate functionalization of nanoparticles allows for targeting of C-type lectin receptors. This family of pattern recognition receptors expressed on innate immune cells, such as macrophages and dendritic cells, can be used to modulate immune responses. In this work, the in vivo safety profile of carbohydrate-functionalized polyanhydride nanoparticles was analyzed following parenteral and intranasal administration in mice. Polyanhydride nanoparticles based on 1,6-bis-(p-carboxyphenoxy)hexane and 1,8-bis-(p-carboxyphenoxy)-3,6-dioxaoctane were used. Nanoparticle functionalization with di-mannose (specifically carboxymethyl-α-D-mannopyranosyl-(1,2)-D-mannopyranoside), galactose (specifically carboxymethyl-β-galactoside), or glycolic acid induced no adverse effects after administration based on histopathological evaluation of liver, kidneys, and lungs. Regardless of the polymer formulation, there was no evidence of hepatic or renal damage or dysfunction observed in serum or urine samples. The histological profile of cellular infiltration and the cellular distribution and kinetics in the lungs of mice administered with nanoparticle treatments followed similar behavior as that observed in the lungs of animals administered with saline. Cytokine and chemokine profiles in bronchoalveolar lavage fluid indicated surface chemistry dependence on modest secretion of IL-6, IP-10, and MCP-1; however, there was no evidence of any deleterious histopathological changes. Based on these analyses, carbohydrate-functionalized nanoparticles are safe for in vivo applications. These results provide foundational information towards the evaluation of the capabilities of these surface-modified nanoparticles as vaccine delivery formulations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D41007
1,6-Dibromohexane, 96%
C6H12Br2
270458
1-Methyl-2-pyrrolidinone, for HPLC, ≥99%
C5H9NO
43729
1-Methyl-2-pyrrolidinone, for metal speciation analysis, ≥99.0% (GC)
C5H9NO
M6762
1-Methyl-2-pyrrolidinone
C5H9NO
242799
1-Methyl-2-pyrrolidinone, spectrophotometric grade, ≥99%
C5H9NO
78769
1-Methyl-2-pyrrolidinone, analytical standard
C5H9NO
691275
Acetic anhydride, Lonza quality, ≥99.5% (GC)
C4H6O3
242845
Acetic anhydride, ACS reagent, ≥98.0%
C4H6O3
320102
Acetic anhydride, ReagentPlus®, ≥99%
C4H6O3
539996
Acetic anhydride, 99.5%
C4H6O3
A6404
Acetic anhydride, reagent grade, ≥98%
C4H6O3
45840
Acetic anhydride
C4H6O3
01-0390
Acetic anhydride, SAJ first grade, ≥93.0%
C4H6O3
01-0380
Acetic anhydride, JIS special grade
C4H6O3
L095004
Acetic anhydride
C4H6O3
E1521
Ethylenediamine, BioXtra
C2H8N2
03550
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
C2H8N2
41008
Ethylenediamine, analytical standard
C2H8N2
391085
Ethylenediamine, purified by redistillation, ≥99.5%
C2H8N2
E26266
Ethylenediamine, ReagentPlus®, ≥99%
C2H8N2
E1649
Ethylenediamine, meets USP testing specifications
C2H8N2
15068
Ethylenediamine solution, technical, 75-80%
C2H8N2
124737
Glycolic acid, ReagentPlus®, 99%
C2H4O3
G8284
Glycolic acid, BioXtra, ≥98.0% (titration)
C2H4O3
94815
Glycolic acid, certified reference material, TraceCERT®
C2H4O3
PHR1427
Glycolic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
C2H4O3
420581
Glycolic acid solution, high purity, 70 wt. % in H2O
C2H4O3
420603
Glycolic acid solution, technical grade, 70 wt. % in H2O
C2H4O3
50610
Glycolic acid solution, technical, ~55% in H2O
C2H4O3
236705
Pentane, anhydrous, ≥99%
C5H12