EMAIL THIS PAGE TO A FRIEND
Natural product research
A new phenylpropanoid from the roots of Euphorbia nematocypha.
PMID 25421575
Abstract
A phytochemical investigation of the ethanolic extracts of the dried roots of Euphorbia nematocypha resulted in the isolation of a new phenylpropanoid, 16-O-caffeoyl-16-hydroxylhexadecanoic acid (1), together with 23 known compounds (2-24). Their structures were elucidated on the basis of spectroscopic data. Compound 1 was first to be isolated as a caffeic acyl long chain alkyl acid from the family Euphorbiaceae. The new isolated phenylpropanoid showed potent cytotoxic activities against the MCF-7 and HeLa cell lines.