EMAIL THIS PAGE TO A FRIEND
Molecular pharmaceutics
Novel Inhaled Combination Powder Containing Amorphous Colistin and Crystalline Rifapentine with Enhanced Antimicrobial Activities against Planktonic Cells and Biofilm of Pseudomonas aeruginosa for Respiratory Infections.
PMID 25423590
Abstract
Colistin has been increasingly used for the treatment of respiratory infections caused by Gram-negative bacteria. Unfortunately parenteral administration of colistin can cause severe adverse effects. This study aimed to develop an inhaled combination dry powder formulation of colistin and rifapentine for the treatment of respiratory infections. The combination formulation was produced by spray-drying rifapentine particles suspended in an aqueous colistin solution. The combination dry powder had enhanced antimicrobial activities against planktonic cells and biofilm cultures of Pseudomonas aeruginosa, with both minimum inhibitory concentration (MIC) and minimum biofilm inhibitory concentration (MBIC) values (2 and 4 mg/L, respectively) being half that of pure colistin (MIC 4 mg/L and MBIC 8 mg/L) and 1/16th that of pure rifapentine (MIC 32 mg/L and MBIC 64 mg/L). High aerosol performance, as measured via an Aerolizer device, was observed with emitted doses>89% and fine particle fraction (FPF) total>76%. The proportion of submicron particles of rifapentine particles was minimized by the attachment of colistin, which increased the overall particle mass and aerodynamic size distribution. Using the spray-drying method described here, stable particles of amorphous colistin and crystalline rifapentine were distributed homogeneously in each stage of the impinger. Unlike the colistin alone formulation, no deterioration in aerosol performance was found for the combination powder when exposed to a high relative humidity of 75%. In our previous study, surface coating by rifampicin contributed to the moisture protection of colistin. Here, a novel approach with a new mechanism was proposed whereby moisture protection was attributed to the carrier effect of elongated crystalline rifapentine particles, which minimized contact between hygroscopic colistin particles. This inhaled combination antibiotic formulation with enhanced aerosol dispersion efficiency and in vitro efficacy could become a superior treatment for respiratory infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0533
Rifapentine
C47H64N4O12
M84445
S-Methylisothiourea hemisulfate salt, 98%
C2H6N2S · 0.5 H2SO4