EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health. Part A
Metal levels in eggs of waterbirds in the New York Harbor (USA): trophic relationships and possible risk to human consumers.
PMID 25424617
Abstract
Health professionals are interested in evaluating the risks that heavy metals pose to eco-receptors and humans. The objective of this study was to examine levels of mercury (Hg), lead (Pb), cadmium (Cd), and other contaminants in waterbirds nesting in the New York harbor in 2012 to determine (1) whether there were species and locational differences, and (2) whether consumption of eggs posed a health risk to predators or humans. For arsenic (As), Pb, Hg, and selenium (Se), species contributed more to variations in levels than location; for Cd and chromium (Cr), location was more significant. Mean metal levels differed among species for all metals, except Cd. Highest levels were As (great black-backed gulls, Larus marinus), Cr (great egret, Ardea alba), Pb (Canada goose, Branta canadensis), and Hg and Se (black-crowned night heron, Nycticorax nycticorax). There were significant locational differences only for herring gulls (Larus argentatus); significant differences were found for all metals. Levels of Hg and Pb may be sufficiently high in eggs of some species to produce adverse effects in predators that eat them. The proportion of samples above 0.3 ppm Hg (U.S. Environmental Protection Agency [EPA] freshwater criteria for freshwater fish), the contaminant of health concern, ranged from 0% (Canada goose, great egret), to 14 and 27% in gulls, to 50% (black-crowned night heron). Some herring gull, great black-backed gull, and black-crowned night heron eggs had 0.5 ppm or higher Hg. Thus, human consumption of eggs may pose a risk to fetuses and young children.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00623
Cadmium, purum p.a., for filling reductors
Cd
265411
Cadmium, foil, thickness 0.5 mm, ≥99.99% trace metals basis
Cd
265330
Cadmium, granular, ≥99%, 5-20 mesh
Cd
414891
Cadmium, granular, 30-80 mesh, ≥99%
Cd
265365
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cd
265454
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Cd
202886
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cd
348600
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Cd
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF17721637
Cadmium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF76768914
Cadmium, foil, 150x150mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF48075453
Cadmium, foil, 195x200mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF94010453
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF90633652
Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cd
GF06836238
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cd
GF71408763
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF78263411
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF84554613
Cadmium, foil, 300x1000mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF19008817
Cadmium, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF20958947
Cadmium, foil, 300x300mm, thickness 0.5mm, as rolled, 99.99%
Cd