EMAIL THIS PAGE TO A FRIEND
mBio
Salmonella evades D-amino acid oxidase to promote infection in neutrophils.
PMID 25425233
Abstract
Neutrophils engulf and kill bacteria using oxidative and nonoxidative mechanisms. Despite robust antimicrobial activity, neutrophils are impaired in directing Salmonella clearance and harbor viable intracellular bacteria during early stages of infection that can subsequently escape to more-permissive cell types. The mechanisms accounting for this immune impairment are not understood. We report that Salmonella limits exposure to oxidative damage elicited by D-amino acid oxidase (DAO) in neutrophils by expressing an ABC importer specific for D-alanine, a DAO substrate found in peptidoglycan stem peptides. A Salmonella dalS mutant defective for D-alanine import was more susceptible to killing by DAO through exposure to greater oxidative stress during infection. This fitness defect was reversed by selective depletion of neutrophils or by inhibition of DAO in vivo with a small-molecule inhibitor. DalS-mediated subversion of neutrophil DAO is a novel host-pathogen interaction that enhances Salmonella survival during systemic infection. Neutrophils engulf Salmonella during early stages of infection, but bacterial killing is incomplete. Very little is known about how Salmonella survives in neutrophils to gain access to other cell types during infection. In this study, we show that D-amino acid oxidase (DAO) in neutrophils consumes D-alanine and that importing this substrate protects Salmonella from oxidative killing by neutrophil DAO. Loss of this importer results in increased bacterial killing in vitro, in neutrophils, and in a mouse model of infection, all phenotypes that are lost upon inhibition of DAO. These findings add mechanistic insight into a novel host-pathogen interaction that has consequences on infection outcome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS017027
6-chlorobenzo[d]isoxazol-3-ol, AldrichCPR
C7H4ClNO2
SML0274
CBIO, ≥95% (HPLC)
C7H4ClNO2
C1200000
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
C11H12Cl2N2O5
31667
Chloramphenicol, VETRANAL, analytical standard
C11H12Cl2N2O5
23276
Chloramphenicol, tested according to Ph.Eur.
C11H12Cl2N2O5
A7377
D-Alanine, ≥98% (HPLC)
C3H7NO2
S4250
D-Serine, ≥98% (TLC)
C3H7NO3
V1255
D-Valine, suitable for cell culture, BioReagent
C5H11NO2
855987
D-Valine, ≥98%
C5H11NO2
53330
DL-Histidine, puriss., ≥99.0% (NT)
C6H9N3O2
H0750000
Histidine, European Pharmacopoeia (EP) Reference Standard
C6H9N3O2
A7469
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, ≥98.5%
C3H7NO2
A4349
L-Alanine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2
05129
L-Alanine, BioUltra, ≥99.5% (NT)
C3H7NO2
44526
L-Alanine, certified reference material, TraceCERT®
C3H7NO2
PHR1110
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2
05130
L-Alanine, ≥99.0% (NT)
C3H7NO2
W381829
L-Alanine, ≥99%
C3H7NO2
A7627
L-Alanine, ≥98% (TLC)
C3H7NO2
492876
L-Alanine-12C3, 99.9 atom % 12C
12C3H7NO2
29469
N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, ≥99.0% (CHN)
C14H26N3O · C7H7O3S
N9877
Nitrilotriacetic acid, Sigma Grade, ≥99%
C6H9NO6
PHR1277
Nitrilotriacetic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C6H9NO6
D9143
o-Dianisidine, peroxidase substrate
C14H16N2O2
33430
o-Dianisidine, for spectrophotometric det. of Au, NO2-, Ce(IV), for the detection of Au, Co, Cu, SCN-, V, ≥97.0%
C14H16N2O2