EMAIL THIS PAGE TO A FRIEND
Medical oncology (Northwood, London, England)
Prodigiosin isolated from cell wall of Serratia marcescens alters expression of apoptosis-related genes and increases apoptosis in colorectal cancer cells.
PMID 25429836
Abstract
Colorectal cancer remains often refractory to classic therapies. In consequence, the search for new anti-tumor agents with minimal toxicity is of particular interest in colon cancer treatment. Prodigiosin as a secondary metabolite of Serratia marcescens induces apoptosis in various kinds of cancer cells with low toxicity on normal cells. In the present study, we evaluated the effect of prodigiosin on proliferation and expression of apoptotic-related genes in HT-29 cells. Malignant cells were treated to various concentrations of prodigiosin and proliferation rate, survivin, Bcl-2, Bax and Bad mRNA levels, caspase 3 activation and apoptosis were evaluated by different cellular and molecular techniques. Treatment of cells with increasing concentration of prodigiosin decreased significantly cell proliferation in a dose- and time-dependent manner. Following 48-h treatment, growth rate was measured to be 77xa0±xa06.8, 41.3xa0±xa03.1 and 46xa0±xa06.3xa0% for 100, 400 and 600xa0nM prodigiosin, respectively, compared to untreated cells. This molecule induced 61.7, 90 and 89xa0% decrease in survivin mRNA level as well as 1.9-, 2.8- and 2.2-fold increase in caspase 3 activation for indicated concentrations of prodigiosin, respectively. The level of Bcl-2 mRNA was inversely proportional to Bax and Bad mRNA levels. Low mRNA levels of Bcl-2 combined with high levels of Bax and Bad mRNAs were correlated to higher apoptosis rate in treated cells. Our data suggest that prodigiosin-induced apoptosis may ascribe to Bcl-2 and survivin inhibition in HT-29 cells and these genes may provide promising molecular targets of prodigiosin. Collectively, prodigiosin may have a great potential for colorectal cancer-directed therapy.