EMAIL THIS PAGE TO A FRIEND
Journal of structural biology
Investigation of resins suitable for the preparation of biological sample for 3-D electron microscopy.
PMID 25433274
Abstract
In the last two decades, the third-dimension has become a focus of attention in electron microscopy to better understand the interactions within subcellular compartments. Initially, transmission electron tomography (TEM tomography) was introduced to image the cell volume in semi-thin sections (∼ 500 nm). With the introduction of the focused ion beam scanning electron microscope, a new tool, FIB-SEM tomography, became available to image much larger volumes. During TEM tomography and FIB-SEM tomography, the resin section is exposed to a high electron/ion dose such that the stability of the resin embedded biological sample becomes an important issue. The shrinkage of a resin section in each dimension, especially in depth, is a well-known phenomenon. To ensure the dimensional integrity of the final volume of the cell, it is important to assess the properties of the different resins and determine the formulation which has the best stability in the electron/ion beam. Here, eight different resin formulations were examined. The effects of radiation damage were evaluated after different times of TEM irradiation. To get additional information on mass-loss and the physical properties of the resins (stiffness and adhesion), the topography of the irradiated areas was analysed with atomic force microscopy (AFM). Further, the behaviour of the resins was analysed after ion milling of the surface of the sample with different ion currents. In conclusion, two resin formulations, Hard Plus and the mixture of Durcupan/Epon, emerged that were considerably less affected and reasonably stable in the electron/ion beam and thus suitable for the 3-D investigation of biological samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90872
Acetone, analytical standard
C3H6O
1006801
Acetone, United States Pharmacopeia (USP) Reference Standard
C3H6O
PHR1219
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O
D2270
Dibutyl phthalate
C16H22O4
152439
Dibutyl phthalate, ReagentPlus®, ≥99%
C16H22O4
18281
Dibutyl phthalate, certified reference material, TraceCERT®
C16H22O4
D1010000
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
C16H22O4
524980
Dibutyl phthalate, 99%
C16H22O4
80100
Dibutyl phthalate, Selectophore
C16H22O4
36736
Dibutyl phthalate, PESTANAL®, analytical standard
C16H22O4
85191
Glutaraldehyde solution, 1.2 % (w/v) glutaraldehyde in H2O
C5H8O2
49630
Glutaraldehyde solution, technical, ~25% in H2O (2.6 M)
C5H8O2
688886
Methyl acetoacetate, Lonza quality, ≥99% (GC)
C5H8O3
10913
Methyl acetoacetate, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C5H8O3
14610
Methyl acetoacetate, analytical standard
C5H8O3
537365
Methyl acetoacetate, ReagentPlus®, 99%
C5H8O3
702587
Potassium ferricyanide(III), powder or chunks, <10 μm, 99%
71649
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
HNa2O4P · 12H2O
71650
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
HNa2O4P · 12H2O
04273
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-101% (calc. to the dried substance)
HNa2O4P · 12H2O
71663
Sodium phosphate dibasic dodecahydrate, tested according to Ph.Eur.
HNa2O4P · 12H2O
30414
Sodium phosphate dibasic dodecahydrate, 99-101% (calc. to the dried substance)
HNa2O4P · 12H2O
14089
Succinic anhydride, puriss., ≥99.0% (NT)
C4H4O3
14090
Succinic anhydride, ≥97.0% (NT)
C4H4O3
239690
Succinic anhydride, ≥99% (GC)
C4H4O3