EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Sesquiterpene lactones from the aerial parts of Vernonia blumeoides growing in Nigeria.
PMID 25433631
Abstract
Four eudesmanolide sesquiterpene lactones (1-4) were isolated from the aerial parts of Vernonia blumeoides used in Nigerian ethnomedicine for the treatment of diarrhea and malaria. Compound 1 demonstrated limited but interesting antibacterial activity against Bacillus, Staphylococcus and Streptococcus species. The crystal structure of 1 allowed the absolute configuration of the stereocentres in the molecule to be assigned.