EMAIL THIS PAGE TO A FRIEND
Cancer letters
The positive is inside the negative: HER2-negative tumors can express the HER2 intracellular domain and present a HER2-positive phenotype.
PMID 25434795
Abstract
Overexpression of human epithelial growth factor receptor 2 (HER2) is a poor prognostic factor in breast cancer. HER2 is a transmembrane receptor comprising an extracellular domain (ECD), a single transmembrane domain, and an intracellular domain (ICD) with tyrosine-kinase activity. Receptor dimerization triggers pivotal effector pathways in cancer, such as phosphatidylinositol 3-kinase (PI3K) signaling. Currently, screening of HER2 in breast tumors for prognostic and therapeutic purposes involves immunohistochemical (IHC) phenotyping for the ECD, in which tumors with IHC scores below 2+ are reported as HER2-negative. We used a label-free liquid chromatography-mass spectrometry (LC-MS) proteomic approach to compare plasma samples from patients with HER2-positive breast tumors and patients with HER2-negative tumors. Patients with HER2-negative tumors expressed higher circulating levels of calpain-10 than patients with HER2-positive tumors. Calpains cleave HER2, releasing its ECD and transforming phenotypically positive tumors into phenotypically negative tumors. Therefore, we investigated the expression of the ICD in HER2-negative samples that overexpressed calpain-10. We found that 16% of HER2-negative tumors were positive for HER2-ICD, which was associated with circulating HER2-ECD. HER2 gene amplification was also observed in some HER2-negative tumors. Positive staining for the PI3K pathway was observed in the HER2-negative, ICD-positive tumors, similar to the HER2-positive cohort. Microarray analysis revealed that HER2-negative, ICD-positive samples clustered between HER2-positive tumors and triple-negative tumors. Survival analysis revealed that outcome in women with HER2-negative, ICD-positive tumors was better than in women bearing HER2-negative, ICD-negative (triple negative) tumors but was quite similar to HER2-positive tumors and worse than women with luminal A tumors. Moreover, in vitro analyses revealed that MDA-MB 231, a triple negative cell line, possesses calpain-10 and HER2-p-ICD up-regulation and blockage of calpain-10 activity promoted an increase in HER2-p-ICD and p-AKT levels, suggesting an increase in these pathways signaling. These data indicate that HER2-negative tumors with HER2-ICD positivity exhibit clinical behavior closer to that of HER2-positive tumors. This indicates a need for HER2-ICD screening when determining the molecular profile of breast tumors. These findings further indicate that lapatinib should be investigated as a target therapy for HER2-ICD-positive breast tumors.