EMAIL THIS PAGE TO A FRIEND
Leukemia research
Non transferrin bound iron (NTBI) in acute leukemias throughout conventional intensive chemotherapy: kinetics of its appearance and potential predictive role in infectious complications.
PMID 25435028
Abstract
We analyzed appearance of non transferrin bound iron (NTBI) in 30 transplant eligible patients with acute myeloid leukemia (AML) and acute lymphoblastic leukemia (ALL) during conventional chemotherapy treatment program and evaluated possible relationship with transfusional body iron intake, iron parameters and clinical complications. For each course, serum samples for NTBI detection were taken prior to chemotherapy, during treatment and during subsequent bone marrow myelosuppression: NTBI was assessed by HPLC. Appearance of NTBI was observed from the start of induction treatment and was still detectable during bone marrow myelosuppression; the recovery of the bone marrow function coincided with the disappearance of NTBI. This kinetic was observed in all subsequent high doses chemotherapy courses, independently from confounding variables such as transfusional iron intake and transferrin saturation. NTBI seems to be a consequence of chemotherapy induced lysis of bone marrow cells and, partly, of hepatocytes after cytotoxic injury. The subsequent persistence of NTBI throughout bone marrow myelosuppression is related to the transient suspension of erythropoietic activity. Moreover, NTBI levels >2μM at the beginning of iatrogenic myelosuppression were associated with higher risk of sepsis caused by Gram negative Bacilli (RR 2.571), also compared with other infectious complications (RR 1.954).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3518
Carbonyl iron, ≥97% Fe basis
Fe
44890
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Fe
12310
Iron, ≥99%, reduced, powder (fine)
Fe
267945
Iron, chips, 99.98% trace metals basis
Fe
338141
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Fe
356808
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Fe
413054
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Fe
513423
Iron, activated, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 99.5% trace metals basis
Fe
IRMM524A
Iron, IRMM® certified Reference Material, 0.1 mm foil
Fe
IRMM524B
Iron, IRMM® certified Reference Material, 0.5 mm wire
Fe
255637
Iron, powder, ≥99.99% trace metals basis
Fe
209309
Iron, powder, −325 mesh, 97%
Fe
267953
Iron, powder, <10 μm, ≥99.9% trace metals basis
Fe
266213
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Fe
266256
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Fe
266248
Iron, wire, diam. 0.5 mm, ≥99.99%
Fe
356824
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Fe
356832
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Fe
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF34071566
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Fe
GF96915134
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Fe
GF35663935
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Fe
GF27511781
Iron, wire reel, 5m, diameter 0.25mm, annealed, 99.99+%
Fe
GF61040023
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Fe