EMAIL THIS PAGE TO A FRIEND
Mutation research. Genetic toxicology and environmental mutagenesis
Genotoxic effects of CdS quantum dots and Ag2S nanoparticles in fish cell lines (RTG-2).
PMID 25435359
Abstract
The increasing use of nanotechnologies will lead to significant releases of engineered nanoparticles into the aquatic environment, where their impact is still poorly characterized. In the present paper, the genotoxic and cytotoxic properties of CdS quantum dots (QDs) and silver sulphide (Ag2S) coated with methyl polyethylene glycol (M-PEG) were investigated in a rainbow trout cell line (RTG-2). The results showed that CdS QDs were highly cytotoxic at high concentrations (10 and 50μg/ml), and exhibited a concentration-dependent genotoxicity in the sub-toxic range (0.01-1μg/ml) after 24h exposure. Ag2S showed neither genotoxic nor cytotoxic effects.