EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
In-syringe dispersive solid-phase extraction using dissolvable layered double oxide hollow spheres as sorbent followed by high-performance liquid chromatography for determination of 11 phenols in river water.
PMID 25435460
Abstract
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed much higher extraction efficiency than normal layered double hydroxides and layered double oxides. The entire extraction process was accomplished in a syringe. After DSPE, the sorbent with the analytes was isolated conveniently by directly expelling the spent sample solution out of the syringe. The analyte-enriched sorbent was then subsequently dissolved by withdrawing an acidic solution into the syringe. The final extract was analyzed by high-performance liquid chromatography with ultraviolet detection. The results showed that this method provided low limits of detection for the phenols (0.005-0.153 μg/L), good linearity (r(2)≥ 0.9956) and relative standard deviations of ≤ 6.7%. The optimized method was applied to water samples from 3 rivers. This simple extraction procedure was demonstrated to be a fast, efficient and convenient DSPE approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32900
Benalaxyl-M, PESTANAL®, analytical standard
C20H23NO3
B6768
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
H3BO3
B7901
Boric acid, for electrophoresis, ≥99.5%
H3BO3
B2645
Boric acid, tablet, 1 g boric acid per tablet
H3BO3
B9645
Boric acid, cell culture tested, plant cell culture tested, ≥99.5%
H3BO3
B1934
Boric acid, Biotechnology Performance Certified, ≥99.5% (titration), Cell Culture Tested
H3BO3
202878
Boric acid, 99.999% trace metals basis
H3BO3
339067
Boric acid, 99.97% trace metals basis
H3BO3
B7660
Boric acid, BioXtra, ≥99.5%
H3BO3
B0394
Boric acid, ACS reagent, ≥99.5%
H3BO3
B0252
Boric acid, ReagentPlus®, ≥99.5%
H3BO3
15663
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
H3BO3
31146
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
H3BO3
11607
Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
H3BO3
426148
Boric acid-11B, ≥99 atom % 11B
H311BO3
M5296
Magnesium nitrate hexahydrate, BioXtra, ≥98%
MgN2O6 · 6H2O
63084
Magnesium nitrate hexahydrate, BioUltra, ≥99.0% (KT)
MgN2O6 · 6H2O
63087
Magnesium nitrate hexahydrate, puriss. p.a., ACS reagent, 98.0-102.0% (KT)
MgN2O6 · 6H2O
203696
Magnesium nitrate hexahydrate, 99.999% trace metals basis
MgN2O6 · 6H2O
237175
Magnesium nitrate hexahydrate, ACS reagent, 99%
MgN2O6 · 6H2O