EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
A quantitiative LC-MS/MS method for the measurement of arachidonic acid, prostanoids, endocannabinoids, N-acylethanolamines and steroids in human plasma.
PMID 25436483
Abstract
Free arachidonic acid is functionally interlinked with different lipid signaling networks including those involving prostanoid pathways, the endocannabinoid system, N-acylethanolamines, as well as steroids. A sensitive and specific LC-MS/MS method for the quantification of arachidonic acid, prostaglandin E2, thromboxane B2, anandamide, 2-arachidonoylglycerol, noladin ether, lineoyl ethanolamide, oleoyl ethanolamide, palmitoyl ethanolamide, steroyl ethanolamide, aldosterone, cortisol, dehydroepiandrosterone, progesterone, and testosterone in human plasma was developed and validated. Analytes were extracted using acetonitrile precipitation followed by solid phase extraction. Separations were performed by UFLC using a C18 column and analyzed on a triple quadrupole MS with electron spray ionization. Analytes were run first in negative mode and, subsequently, in positive mode in two independent LC-MS/MS runs. For each analyte, two MRM transitions were collected in order to confirm identity. All analytes showed good linearity over the investigated concentration range (r>0.98). Validated LLOQs ranged from 0.1 to 190ng/mL and LODs ranged from 0.04 to 12.3ng/mL. Our data show that this LC-MS/MS method is suitable for the quantification of a diverse set of bioactive lipids in plasma from human donors (n=32). The determined plasma levels are in agreement with the literature, thus providing a versatile method to explore pathophysiological processes in which changes of these lipids are implicated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2016
3α,21-Dihydroxy-5α-pregnan-20-one, ≥95%
C21H34O3
P8887
5α-Pregnan-3α-ol-20-one, solid
C21H34O2
A9477
Aldosterone, ≥95% (HPLC)
C21H28O5
A1542
Ammonium acetate, for molecular biology, ≥98%
C2H7NO2
A7330
Ammonium acetate, BioXtra, ≥98%
C2H7NO2
09689
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
C2H7NO2
A7262
Ammonium acetate, reagent grade, ≥98%
C2H7NO2
73594
Ammonium acetate, for mass spectrometry, eluent additive for LC-MS
C2H7NO2
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2
09691
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
C2H7NO2
31579
Androsterone, VETRANAL, analytical standard
C19H30O2
A9673
Arachidonic acid, from porcine liver, ≥99% (GC), liquid
C20H32O2
A3555
Arachidonic acid, from porcine liver, BioReagent, suitable for cell culture, ≥99% (GC)
C20H32O2
A3925
Arachidonic acid, from porcine liver, ≥85% (capillary GC), liquid
C20H32O2
10931
Arachidonic acid, >95.0% (GC)
C20H32O2
A3611
Arachidonic acid, from non-animal source, ≥98.5% (GC)
C20H32O2
A8848
Arachidonoyl dopamine, ≥98% (HPLC), ethanol solution
C28H41NO3
C8115
Coronatine, from Pseudomonas syringae pv Glycinea, >95% (HPLC)
C18H25NO4
D2250000
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
C20H32O5
07220
DL-2-Aminobutyric acid, puriss., ≥99.0% (NT)
C4H9NO2
162663
DL-2-Aminobutyric acid, ReagentPlus®, 99%
C4H9NO2
S5386
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
C3H7NO3
S4375
DL-Serine, ≥98% (TLC)
C3H7NO3
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
437549
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
324027
Ethyl acetate, PRA grade, ≥99.5%
C4H8O2
58958
Ethyl acetate, analytical standard
C4H8O2
17023
Ethyl acetate, tested according to Ph.Eur.
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2