EMAIL THIS PAGE TO A FRIEND
Epigenetics
Non-referenced genome assembly from epigenomic short-read data.
PMID 25437048
Abstract
Current computational methods used to analyze changes in DNA methylation and chromatin modification rely on sequenced genomes. Here we describe a pipeline for the detection of these changes from short-read sequence data that does not require a reference genome. Open source software packages were used for sequence assembly, alignment, and measurement of differential enrichment. The method was evaluated by comparing results with reference-based results showing a strong correlation between chromatin modification and gene expression. We then used our de novo sequence assembly to build the DNA methylation profile for the non-referenced Psammomys obesus genome. The pipeline described uses open source software for fast annotation and visualization of unreferenced genomic regions from short-read data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5760
4-(4-Methoxybenzylamino)-7-nitrobenzofurazan
C14H12N4O4