EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Design and synthesis of sulfonamide derivatives of pyrrolidine and piperidine as anti-diabetic agents.
PMID 25437620
Abstract
Type 2 diabetes (T2D) is a lifestyle disease affecting millions of people worldwide. Various therapies are available for the management of T2D and dipeptidyl peptidase-IV (DPP-IV) inhibition has emerged as a promising therapy for the treatment of type 2 diabetes (T2D). Here we report design, synthesis and in vitro efficacy of sulfonamide derivatives of pyrrolidine and piperidine as anti-diabetic agents. Amongst all the compounds synthesized in this series, 9a, is the most potent (IC50 = 41.17 nM).