EMAIL THIS PAGE TO A FRIEND
Cellular reprogramming
β-Cell differentiation of human pancreatic duct-derived cells after in vitro expansion.
PMID 25437872
Abstract
β-Cell replacement therapy is a promising field of research that is currently evaluating new sources of cells for clinical use. Pancreatic epithelial cells are potent candidates for β-cell engineering, but their large-scale expansion has not been evidenced yet. Here we describe the efficient expansion and β-cell differentiation of purified human pancreatic duct cells (DCs). When cultured in endothelial growth-promoting media, purified CA19-9(+) cells proliferated extensively and achieved up to 22 population doublings over nine passages. While proliferating, human pancreatic duct-derived cells (HDDCs) downregulated most DC markers, but they retained low CK19 and SOX9 gene expression. HDDCs acquired mesenchymal features but differed from fibroblasts or pancreatic stromal cells. Coexpression of duct and mesenchymal markers suggested that HDDCs were derived from DCs via a partial epithelial-to-mesenchymal transition (EMT). This was supported by the blockade of HDDC appearance in CA19-9(+) cell cultures after incubation with the EMT inhibitor A83-01. After a differentiation protocol mimicking pancreatic development, HDDC populations contained about 2% of immature insulin-producing cells and showed glucose-unresponsive insulin secretion. Downregulation of the mesenchymal phenotype improved β-cell gene expression profile of differentiated HDDCs without affecting insulin protein expression and secretion. We show that pancreatic ducts represent a new source for engineering large amounts of β-like-cells with potential for treating diabetes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6273
2-Propylpentanoic acid
C8H16O2
93049
Forskolin, analytical standard
C22H34O7
I7378
Indomethacin, ≥99% (TLC)
C19H16ClNO4
I8280
Indomethacin, meets USP testing specifications
C19H16ClNO4
1341001
Indomethacin, United States Pharmacopeia (USP) Reference Standard
C19H16ClNO4
I0200000
Indomethacin, European Pharmacopoeia (EP) Reference Standard
C19H16ClNO4
PHR1247
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
C19H16ClNO4
N5535
Niacinamide, meets USP testing specifications
C6H6N2O
PHR1033
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
C6H6N2O
1462006
Niacinamide, United States Pharmacopeia (USP) Reference Standard
C6H6N2O
72340
Nicotinamide, ≥99.5% (HPLC)
C6H6N2O
N3376
Nicotinamide, ≥98% (HPLC), powder
C6H6N2O
N0636
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H6N2O
72345
Nicotinamide, ≥98.5% (HPLC)
C6H6N2O
72347
Nicotinamide, tested according to Ph.Eur.
C6H6N2O
N0600000
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
C6H6N2O
PHR1061
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C8H16O2
1708707
Valproic acid, United States Pharmacopeia (USP) Reference Standard
C8H16O2
V0033000
Valproic acid, European Pharmacopoeia (EP) Reference Standard
C8H16O2
Y0001422
Valproic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H16O2