EMAIL THIS PAGE TO A FRIEND
International journal of surgery (London, England)
Effects of lazaroid U-74389G on intestinal ischemia and reperfusion injury in porcine experimental model.
PMID 25438077
Abstract
The readmission of molecular oxygen into an ischemic tissue promotes the oxidation of resuscitated tissue with certain pathophysiologic mechanisms. Twenty four pigs (male or female) were randomized in this study. The animals were allocated to four groups with an equal number (n = 6) in each group: (1) control group-ischemia for 30 min and reperfusion for 60 min. (2) control group-ischemia for 30 min and reperfusion for 120 min. (3) ischemia for 30 min and immediate iv injection of lazaroid U-74389G and reperfusion for 60 min. (4) ischemia for 30 min and immediate iv injection of lazaroid U-74389G and reperfusion for 120 min. We investigated further the role of an antioxidant molecule such as U-74389G and we concluded that there is statistically significant relation in MDA (malondialdeyde), TNF -α (tumor necrosis factor-α) measurement in tissue, while the histological score in the groups that the lazaroid was administered was improved. In many emergency clinical situations, such as reperfusion of the intestine, the role of U-74389G can be protective.