EMAIL THIS PAGE TO A FRIEND
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons
Are muscle relaxants needed for nasal intubation in propofol and remifentanil anesthesia?
PMID 25438272
Abstract
The authors hypothesized that a muscle relaxant would have no meaningful difference in intubation conditions during nasal intubation under remifentanil and propofol anesthesia. This parallel-group, double-blinded, randomized controlled trial included 44 patients who received saline (S group; n = 22) or rocuronium (R group; n = 22). In addition to remifentanil 0.5 μg/kg per minute and propofol 5 mg/kg per hour, propofol 0.5 mg/kg was administered until loss of consciousness. Nasal intubation was performed 10 minutes after administration of R or S 0.6 mg/kg. Significant differences in intubation conditions and salivary amylase levels before and after intubation were tested (P < .05). Vocal cord status (P = .003) and response to intubation or cuff filling (P = .008) were significantly different, but intubation conditions were not. Salivary amylase level was significantly lower with R administration (P = .022). No patient complained of postoperative throat pain and hoarseness. Muscle relaxants during nasal intubation performed after bolus administration of propofol 0.9 mg/kg in addition to 10 minutes of remifentanil 0.5 μg/kg per minute plus propofol 5 mg/kg per hour are unnecessary.