EMAIL THIS PAGE TO A FRIEND
The Israel Medical Association journal : IMAJ
Harmonization of autoimmune diagnostics with antinuclear antibody testing algorithm: approach of appropriateness and clinical relevance.
PMID 25438456