EMAIL THIS PAGE TO A FRIEND
Ultrasound in medicine & biology
Usefulness of real-time elastography strain ratio in the assessment of bile duct ligation-induced liver injury and the hepatoprotective effect of chitosan: an experimental animal study.
PMID 25438858
Abstract
The purpose of the study described here was to evaluate the usefulness of the elastographic strain ratio in the assessment of liver changes in an experimental animal setting and the hepatoprotective effects of chitosan. Ultrasonography and Strain Ratio calculation were performed before and after bile duct ligation (BDL) in three groups of Wistar albino rats (n = 10 animals per group): (i) rats subjected to bile duct ligation only; (ii) rats subjected to bile duct ligation and administered chitosan for 14 d; (iii) rats subjected to bile duct ligation and administered chitosan for 7 d. The results were compared with the laboratory data and pathologic findings. Strain ratios revealed an increase in liver stiffness after bile duct ligation (p < 0.05), except in the group with chitosan administered for 7 d, and agreed with laboratory and pathology data. In conclusion, strain ratio can be used as an experimental research instrument in the assessment of liver response to injury. To the best of our knowledge, this is the first study reporting on the usefulness of the sonoelastographic liver-to-kidney strain ratio in assessing the effects of experimentally induced liver lesions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E5134
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, for molecular biology, 99.0-101.0% (titration)
C10H14N2Na2O8 · 2H2O
E1644
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E6635
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, plant cell culture tested, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E5513
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E4884
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
ED2SS
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
C10H14N2Na2O8 · 2H2O
03677
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
C10H14N2Na2O8 · 2H2O
27285
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
PHR1068
Ethylenediaminetetraacetic acid disodium salt dihydrate
C10H14N2Na2O8 · 2H2O
03685
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
C10H14N2Na2O8 · 2H2O
2854
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
C10H14N2Na2O8
E7889
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
C10H14N2Na2O8