EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Atmospheric pressure gas chromatography-time-of-flight-mass spectrometry (APGC-ToF-MS) for the determination of regulated and emerging contaminants in aqueous samples after stir bar sorptive extraction (SBSE).
PMID 25440658
Abstract
This work presents the development, optimization and validation of a multi-residue method for the simultaneous determination of 102 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), and several types of pesticides in aqueous matrices. Water samples were processed using stir bar sorptive extraction (SBSE) after the optimization of several parameters: agitation time, ionic strength, presence of organic modifiers, pH, and volume of the derivatizing agent. Target compounds were extracted from the bars by liquid desorption (LD). Separation, identification and quantification of analytes were carried out by gas chromatography (GC) coupled to time-of-flight (ToF-MS) mass spectrometry. A new ionization source, atmospheric pressure gas chromatography (APGC), was tested. The optimized protocol showed acceptable recovery percentages (50-100%) and limits of detection below 1ngL(-1) for most of the compounds. Occurrence of 21 out of 102 analytes was confirmed in several environmental aquatic matrices, including seawater, sewage effluent, river water and groundwater. Non-target compounds such as organophosphorus flame retardants were also identified in real samples by accurate mass measurement of their molecular ions using GC-APGC-ToF-MS. To the best of our knowledge, this is the first time that this technique has been applied for the analysis of contaminants in aquatic systems. By employing lower energy than the more widely used electron impact ionization (EI), AGPC provides significant advantages over EI for those substances very susceptible to high fragmentation (e.g., fragrances, pyrethroids).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
415820
2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, 97%
C24H27NO2
437395
2-Ethylhexyl 4-(dimethylamino)benzoate, 98%
C17H27NO2
78848
2-Ethylhexyl 4-methoxycinnamate, 98%
C18H26O3
55529
2-Ethylhexyl 4-methoxycinnamate, analytical standard
C18H26O3
W514500
2-Ethylhexyl salicylate, ≥99%
C15H22O3
52184
2-Ethylhexyl salicylate, analytical standard
C15H22O3
437379
2-Ethylhexyl salicylate, 99%
C15H22O3
01479
2-Hydroxy-4-methoxybenzophenone, certified reference material, TraceCERT®
C14H12O3
H36206
2-Hydroxy-4-methoxybenzophenone, 98%
C14H12O3
103160
2-Hydroxybenzophenone, 99%
C13H10O2
78551
3-(4-Methylbenzylidene)camphor, ≥98.0% (GC)
C18H22O
61547
3-(4-Methylbenzylidene)camphor, certified reference material, TraceCERT®
C18H22O
220434
3-Hydroxybenzophenone, 99%
C13H10O2
442840
4,4′-Isopropylidenediphenol, analytical standard
C15H16O2
H20202
4-Hydroxybenzophenone, 98%
C13H10O2
524190
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 97%
C12H7Cl3O2
45806
α-Cypermethrin, PESTANAL®, analytical standard
C22H19Cl2NO3
16633
Avobenzone, analytical standard
C20H22O3
1045337
Avobenzone, United States Pharmacopeia (USP) Reference Standard
C20H22O3
PHR1073
Avobenzone, Pharmaceutical Secondary Standard; Certified Reference Material
C20H22O3
46003
β-Cyfluthrin, PESTANAL®, analytical standard
C22H18Cl2FNO3
33738
Baythroid®, PESTANAL®, analytical standard
C22H18Cl2FNO3
471178
Benzophenone-d10, 99 atom % D
C13D10O
84260
Benzyl salicylate, purum, ≥99.0% (GC)
C14H12O3
112380
Benzyl salicylate, 98%
C14H12O3
W215104
Benzyl salicylate, ≥98%, FG
C14H12O3
51031
Benzyl salicylate, analytical standard
C14H12O3
N11203
Bifenthrin, analytical standard
C23H22ClF3O2
239658
Bisphenol A, ≥99%
C15H16O2
42088
Bisphenol A, certified reference material, TraceCERT®
C15H16O2