EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Salt-independent hydrophobic displacement chromatography for antibody purification using cyclodextrin as supermolecular displacer.
PMID 25441076
Abstract
Hydrophobic interaction chromatography (HIC) offers an orthogonal selectivity to ion exchange chromatography and the combination of the two processes can provide a potential cost-effective alternative to protein A chromatography in industrial antibody purification. However, the application of HIC is limited by its close dependence on high concentrations of kosmotropic salts to achieve desired separation. These salts can cause antibody precipitation and induce the corrosion of manufacturing facilities. Here, we report a new strategy of salt-independent HIC, which can capture antibody at the physiological salt concentration and allow the recovery of bound proteins through cyclodextrin (CD)-based displacement elution. Hydrophobicity-intensified HIC media with different coupling amount of phenyl ligands were prepared and assessed for their antibody binding capacity and selectivity. β-CD was investigated for its supermolecular interaction with phenyl ligands and elution capacity as a displacer. The results clarified a nearly linear correlation between binding capacity of human immunoglobulin G (IgG) and phenyl coupling density in the range of 44-159 μmol/mL. The host-guest interaction between β-CD and the phenyl ligands revealed a modest binding strength (Ka=4.1×10(3) M(-1)), and 15 mM β-CD solution showed a general effectiveness as displacement eluent for these HIC media, with IgG recovery varying with the ligand density. This strategy allowed the direct purification of human IgG from serum with satisfactory purity. The whole procedure of this method, including loading and elution, can be performed under physiological conditions. We expect such a salt-independent mode of HIC could be used as a capture or intermediate step in industrial antibody purification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
Y0001470
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
8S61336
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
C7H6O2