EMAIL THIS PAGE TO A FRIEND
Animal science journal = Nihon chikusan Gakkaiho
A new rolling culture-based in vitro fertilization system capable of reducing polyspermy in porcine oocytes.
PMID 25441619
Abstract
The high incidence of polyspermy is one of the major obstacles during in vitro fertilization (IVF) in pigs. To overcome this, we developed a novel IVF method, which involves constant rotation. Oocytes matured in vitro were mixed with spermatozoa (0.2 × 10(5) sperm/mL) in an IVF medium (200 μL) using a 200 μL PCR tube. This tube was then rotated at 1 rpm for 6 h at 38.5°C in a rotation mixer (experimental group). A second PCR tube was simultaneously cultured without rotation (control group). The rate of polyspermy was evaluated 12 h after insemination and was significantly (P < 0.05; 21.0% vs. 48.3%) lower in the experimental group than in the control group. Sperm penetration rate was similar in oocytes from the experimental and control groups (75.2% vs. 83.1%). However, monospermic fertilization rate of the oocytes was significantly (P < 0.05; 44.8% vs. 21.2%) higher in the experimental group than in the control group. Furthermore, the rate of blastocyst formation (30.1% vs. 20.8%) increased in the experimental group, as compared to the control group. This present system will contribute to increase the efficacy of blastocyst production through reduction of polyspermic penetration.