EMAIL THIS PAGE TO A FRIEND
International journal of radiation oncology, biology, physics
Neoadjuvant sandwich treatment with oxaliplatin and capecitabine administered prior to, concurrently with, and following radiation therapy in locally advanced rectal cancer: a prospective phase 2 trial.
PMID 25442042
Abstract
Systemic failure remains the major challenge in management of locally advanced rectal cancer (LARC). To optimize the timing of neoadjuvant treatment and enhance systemic control, we initiated a phase 2 trial to evaluate a new strategy of neoadjuvant sandwich treatment, integrating induction chemotherapy, concurrent chemoradiation therapy, and consolidation chemotherapy. Here, we present preliminary results of this trial, reporting the tumor response, toxicities, and surgical complications. Fifty-one patients with LARC were enrolled, among which were two patients who were ineligible because of distant metastases before treatment. Patients were treated first with one cycle of induction chemotherapy consisting of oxaliplatin, 130 mg/m² on day 1, with capecitabine, 1000 mg/m² twice daily for 14 days every 3 weeks (the XELOX regimen), followed by chemoradiation therapy, 50 Gy over 5 weeks, with the modified XELOX regimen (oxaliplatin 100 mg/m²), and then with another cycle of consolidation chemotherapy with the XELOX regimen. Surgery was performed 6 to 8 weeks after completion of radiation therapy. Tumor responses, toxicities, and surgical complications were recorded. All but one patent completed the planned schedule of neoadjuvant sandwich treatment. Neither life-threatening blood count decrease nor febrile neutropenia were observed. Forty-five patents underwent optimal surgery with total mesorectal excision (TME). Four patients refused surgery because of clinically complete response. There was no perioperative mortality in this cohort. Five patients (11.1%) developed postoperative complications. Among the 45 patients who underwent TME, pathologic complete response (pCR), pCR or major regression, and at least moderate regression were achieved in 19 (42.2%), 37 (82.2%), and 44 patients (97.8%), respectively. Preliminary results suggest that the strategy of neoadjuvant sandwich treatment using XELOX regimen as induction, concomitant, and consolidation chemotherapy to the conventional radiation is well tolerated. The strategy is highly effective in terms of pCR and major regression, which warrants further investigation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3897
2′-Deoxycytidine, ≥99% (HPLC)
C9H13N3O4