EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Astrocytes show reduced support of motor neurons with aging that is accelerated in a rodent model of ALS.
PMID 25443290
Abstract
Astrocytes play a crucial role in supporting motor neurons in health and disease. However, there have been few attempts to understand how aging may influence this effect. Here, we report that rat astrocytes show an age-dependent senescence phenotype and a significant reduction in their ability to support motor neurons. In a rodent model of familial amyotrophic lateral sclerosis (ALS) overexpressing mutant superoxide dismutase 1 (SOD1), the rate of astrocytes acquiring a senescent phenotype is accelerated and they subsequently provide less support to motor neurons. This can be partially reversed by glial cell line-derived neurotrophic factor (GDNF). Replacing aging astrocytes with young ones producing GDNF may therefore have a significant survival promoting affect on aging motor neurons and those lost through diseases such as ALS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S100
Mianserin hydrochloride, analytical standard, for drug analysis
C18H20N2 · HCl