EMAIL THIS PAGE TO A FRIEND
Pharmacological reports : PR
Rupatadine effectively prevents the histamine-induced up regulation of histamine H1R and bradykinin B2R receptor gene expression in the rat paw.
PMID 25443720
Abstract
Activation of histamine H1 receptor (H1R) is a well-known hallmark of allergic and inflammatory pathology. Both types of bradykinin receptors (B1R and B2R) are also known to contribute significantly to the latter and some sort of functional interaction between them and H1R has been alluded to in the past. Here we use an experimental model of rat paw oedema formation to examine the effect of exogenously added histamine on the gene expression of H1R and bradykinin receptors B1R and B2R, alone or in combination to rupatadine, a second generation antihistamine agent. Histamine-induced oedema formation was monitored with a plethysmometer. The gene expression of H1R, B1R and B2R was analyzed with both conventional and real-time PCR. Rupatadine fumarate was used in pure form and administered intraperitoneally, prior to histamine injection into the paw. Microscopy of haematoxylin and eosin-stained sections of paw tissue was used to examine effects on tissue architecture. Histamine injection into the paw resulted in significant up regulation of H1R and B2R without inducing significant cellular infiltration, but appears to affect less the expression of B1R. Rupatadine was, under the conditions used in this study, very effective in preventing this effect and in suppressing oedema formation through its antihistamine action. Rupatadine has a suppressing effect on H1R and B2R gene expression which could add to its efficacy towards allergy and allergy-like conditions.