EMAIL THIS PAGE TO A FRIEND
Journal of contaminant hydrology
Evaluation of leaching potential of three systemic neonicotinoid insecticides in vineyard soil.
PMID 25444119
Abstract
Dinotefuran (DNT), imidacloprid (IMD), and thiamethoxam (THM) are commonly used neonicotinoid insecticides in a variety of agriculture operations. Although these insecticides help growers control pest infestation, the residual environmental occurrence of insecticides may cause unintended adverse ecological consequences to non-target species. In this study, the leaching behavior of DNT, IMD, and THM was investigated in soils collected from an active AgriLife Research Extension Center (AREC) vineyard. A series of column experiments were conducted to evaluate the leaching potential of insecticides under two experimental scenarios: a) individual pulse mode, and b) mixed pulse mode. In both scenarios, the breakthrough pattern of the insecticides in the mostly acidic to neutral vineyard soil clearly demonstrates medium to high leachability. Of the three insecticides studied for leaching, DNT has exhibited high leaching potential and exited the column with fewer pore volumes, whereas IMD was retained for longer, indicating lower leachability. Relative differences in leaching behavior of neonicotinoids could be attributed to their solubility with the leaching pattern IMD