EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
HDL cholesterol performance using an ultracentrifugation reference measurement procedure and the designated comparison method.
PMID 25444739
Abstract
Accurate high-density lipoprotein cholesterol (HDL-C) measurements are important for management of cardiovascular diseases. The US Centers for Disease Control and Prevention (CDC) and Cholesterol Reference Method Laboratory Network (CRMLN) perform ultracentrifugation (UC) reference measurement procedure (RMP) to value assign HDL-C. Japanese CRMLN laboratory (Osaka) concurrently runs UC procedure and the designated comparison method (DCM). Osaka performance of UC and DCM was examined and compared with CDC RMP. CDC RMP involved UC, heparin-MnCl₂ precipitation, and cholesterol analysis. CRMLN DCM for samples containing <200 mg/dl triglycerides involved 50-kDa dextran sulfate-MgCl2 precipitation and cholesterol determination. HDL-C regression equations obtained with CDC (x) and Osaka (y) were y=0.992x+0.542 (R(2)=0.996) for Osaka UC and y=1.004x-0.181 (R(2)=0.998) for DCM. Pass rates within ±1 mg/dl of the CDC target value were 91.9 and 92.1% for Osaka UC and DCM, respectively. Biases at 40 mg/dl HDL-C were +0.22 and -0.02 mg/dl for Osaka UC and DCM, respectively. Osaka UC and DCM were highly accurate, precise, and stable for many years, assisting manufacturers to calibrate products for clinical laboratories to accurately measure HDL-C for patients, calculate non-HDL-C, and estimate low-density lipoprotein cholesterol with the Friedewald equation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
GF74177736
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF99005193
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF90559794
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF03428625
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF31519975
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF60677646
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF64815239
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF41418378
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF33640860
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF85367605
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn
GF87981564
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mn
GF53022833
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mn
GF71274258
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Mn
GF70055524
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF86610539
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF00832081
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF61911288
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF94645233
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF05710430
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF27632055
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF11021094
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mn
GF34573697
Manganese, rod, 35mm, diameter 2.0mm, cast, 99.5%
Mn
GF38495885
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mn
GF42039918
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF87355334
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF15798730
Manganese, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn