EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Safety assessment of dietary bamboo charcoal powder: a 90-day subchronic oral toxicity and mutagenicity studies.
PMID 25445512
Abstract
Vegetable carbon has been used as food additive in EU (E153) and China for many years; however, no experimental data have been available on its dietary safety. This study was designed to evaluate the subchronic toxicity and genotoxicity of bamboo charcoal powder (BCP). In the study of subchronic oral toxicity, BCP was administered orally at doses of 2.81, 5.62, and 11.24 g/kg BW for 90 days to SD rats. Additional satellite groups from the control group and high dose group were observed for a 28-day recovery period. At the end of the treatment and recovery periods, animals were sacrificed, and their organs were weighed and blood samples were collected. The toxicological endpoints observed included clinical signs, food consumption, body and organ weights, hematological and biochemical parameters, macroscopic and microscopic examinations. The results showed no significant differences between the BCP treated groups and control group. The genotoxicity of BCP was assessed with the Salmonella typhimurium mutagenicity assay (Ames test) and a combination of comet assay and mammalian erythrocyte micronucleus protocol. The results did not reveal any genotoxicity of BCP. Based on our study, the no-observed-adverse-effect level (NOAEL) for BCP is 11.24 g/kg BW/day.