EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Risk assessment derived from migrants identified in several adhesives commonly used in food contact materials.
PMID 25445514
Abstract
Adhesives are used to manufacture multilayer materials, where their components pass through the layers and migrate to the food. Nine different adhesives (acrylic, vinyl and hotmelt) and their migration in 21 laminates for future use as market samples have been evaluated and risk assessment has been carried out. A total of 75 volatiles and non volatile compounds were identified by gas chromatography-mass spectrometry and ultra-performance liquid chromatography coupled to quadrupole time-of-flight mass spectrometry. Most of the compounds migrated below their specific migration limit (SML), lowest observed adverse effect level (LOAEL), no observed adverse effect level (NOAEL) and values recommended by Cramer. Six compounds classified as high toxicity class III according to Cramer classification, migrated over their SML and exposure values recommended by Cramer, when they were applied in the full area of the packaging. Nevertheless, these adhesives fulfill the threshold in the real application as they are applied in a small area of the packaging.