EMAIL THIS PAGE TO A FRIEND
Journal of drug education
Alcohol consumption and negative sex-related consequences among college women: the moderating role of alcohol protective behavioral strategies.
PMID 25445810
Abstract
Alcohol protective behavioral strategies (PBS) limit overall negative consequences; however, less is known about the relationship between PBS and negative sex-related consequences. The purpose of the current study was to examine the moderating effects of 2 distinct types of PBS-controlled consumption strategies and serious harm reduction strategies-on the relationship between alcohol consumption and alcohol-related risky sexual behavior and sexual victimization. Participants were 459 undergraduate women (ages 18-25) who had consumed alcohol within the past 30 days. Both types of PBS significantly qualified the alcohol-sexual victimization link, but neither type of PBS qualified the alcohol-risky sexual behavior link.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D60400
2,3-Dichloro-5,6-dicyano-p-benzoquinone, 98%
C8Cl2N2O2