EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Novel in situ gelling ocular films for the opioid growth factor-receptor antagonist-naltrexone hydrochloride: fabrication, mechanical properties, mucoadhesion, tolerability and stability studies.
PMID 25445974
Abstract
Naltrexone hydrochloride (NTX) is an innovative drug used in ophthalmology for treatment of ocular surface diseases such as impaired corneal wound healing and severe dry eye. Poor chemical stability has been a major limitation for development of NTX in solution form. The aim of this study was to develop and characterise NTX in situ ocular films for enhanced chemical stability and improved ocular tolerability. The films were prepared from different amorphous polymers and characterised for physicochemical compatibility, moisture-sorption, surface pH, mechanical properties, sterilisability, surface morphology, mucoadhesion, in vitro release, conjunctival irritation and accelerated stability at 40°C/75% relative humidity for 3 months. Glycerin (GLY)-plasticised films exhibited significantly better mechanical properties, compared with polyethylene glycol (PEG) 400 and triethylcitrate (TEC)-plasticised formulations. Superior mucoadhesion was recorded for F7 and F9 plasticised with GLY and PEG 400, respectively. The stability of NTX was significantly enhanced more than 18-times, compared with the solution form. Combination of carboxymethylcellulose sodium (CMC) and sodium alginate (ALG) in a film formulation demonstrated minimal % moisture sorption, good mechanical properties, in vitro release, excellent chemical stability and minimal conjunctival irritation lending them as promising ocular formulations.