EMAIL THIS PAGE TO A FRIEND
Neurotoxicology and teratology
Angiotensin AT2 receptor stimulation inhibits activation of NADPH oxidase and ameliorates oxidative stress in rotenone model of Parkinson's disease in CATH.a cells.
PMID 25446015
Abstract
Oxidative stress has long been considered as a major contributing factor in the pathogenesis of Parkinson's disease (PD). The brain has an independent local renin-angiotensin system (RAS). Angiotensin II (Ang II) activates NADPH-dependent oxidases, which are a major source of superoxide and are upregulated in major aging-related diseases such as hypertension and neurodegenerative disease. In this study, we firstly examined whether CGP42112, an AT2 receptor (AT2R) agonist, may exert direct protective effects on the rotenone-induced CATH.a cell injury in vitro. We used CATH.a cell line to evaluate changes in cultured dopaminergic neuron levels of superoxide dismutase (SOD), glutathione (GSH) and reactive oxygen species (ROS). We also evaluated expression of NADPH oxidase, AT1 and AT2 receptors in treated with phosphate buffer saline (PBS), rotenone, Ang II, AT2R agonist CGP42112, or AT2R antagonist PD123319, alone and combined (n=6, each group). Quantitative reverse transcriptase PCR (qRT-PCR) and western blot were used to determine messenger RNA (mRNA) and protein levels of the AT1, AT2 receptors and NADPH oxidase. ROS generation was determined by the dichlorodihydrofluorescein diacetate fluorescent probe assay. The levels of SOD and GSH were measured by using available kits. In our study, CGP42112 (100nM) significantly reduced rotenone-induced oxidative stress and elevated the total SOD activity and GSH level. In addition, CGP42112 significantly increased AT2R expression and attenuated Ang II-induced NADPH oxidase activation, and these effects were completely abolished by the AT2R antagonist, PD123319 (1μM). Our results suggest that CGP42112 attenuates rotenone-induced oxidative stress in CATH.a neuron via activating AT2R and suppressing NADPH oxidase expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45656
Rotenone, PESTANAL®, analytical standard
C23H22O6