EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Treatment with insulin-like growth factor 1 receptor inhibitor reverses hypoxia-induced epithelial-mesenchymal transition in non-small cell lung cancer.
PMID 25446090
Abstract
Insulin-like growth factor 1 receptor (IGF1R) is expressed in many types of solid tumors including non-small cell lung cancer (NSCLC), and enhanced activation of IGF1R is thought to reflect cancer progression. Epithelial-mesenchymal transition (EMT) has been established as one of the mechanisms responsible for cancer progression and metastasis, and microenvironment conditions, such as hypoxia, have been shown to induce EMT. The purposes of this study were to address the role of IGF1R activation in hypoxia-induced EMT in NSCLC and to determine whether inhibition of IGF1R might reverse hypoxia-induced EMT. Human NSCLC cell lines A549 and HCC2935 were exposed to hypoxia to investigate the expression of EMT-related genes and phenotypes. Gene expression analysis was performed by quantitative real-time PCR and cell phenotypes were studied by morphology assessment, scratch wound assay, and immunofluorescence. Hypoxia-exposed cells exhibited a spindle-shaped morphology with increased cell motility reminiscent of EMT, and demonstrated the loss of E-cadherin and increased expression of fibronectin and vimentin. Hypoxia also led to increased expression of IGF1, IGF binding protein-3 (IGFBP3), and IGF1R, but not transforming growth factor β1 (TGFβ1). Inhibition of hypoxia-inducible factor 1α (HIF1α) with YC-1 abrogated activation of IGF1R, and reduced IGF1 and IGFBP3 expression in hypoxic cells. Furthermore, inhibition of IGF1R using AEW541 in hypoxic condition restored E-cadherin expression, and reduced expression of fibronectin and vimentin. Finally, IGF1 stimulation of normoxic cells induced EMT. Our findings indicated that hypoxia induced EMT in NSCLC cells through activation of IGF1R, and that IGF1R inhibition reversed these phenomena. These results suggest a potential role for targeting IGF1R in the prevention of hypoxia-induced cancer progression and metastasis mediated by EMT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y102
YC-1, powder
C19H16N2O2