EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Pentraxin-3 and VEGF in POEMS syndrome: a 2-year longitudinal study.
PMID 25447599
Abstract
Circulating Pentraxin 3 (PTX3) and vascular endothelial growth factor (VEGF) levels were measured longitudinally (mean follow-up 2 years) by ELISA in 6 patients with polyneuropathy, organomegaly, endocrinopathy, M-protein, skin changes (POEMS) syndrome and 16 controls. Expression of PTX3 was also assessed (immunohistochemistry) on sural nerve biopsies from POEMS and vasculitic neuropathy patients. No correlation was found between PTX3 and VEGF levels in POEMS or controls. Sural nerve biopsies from vasculitic neuropathy patients, but not those from POEMS syndrome, showed strong PTX3 staining. PTX3, expression of vessel inflammation/remodeling, and VEGF, crucial pro-angiogenic cytokine, appear to be independently regulated in POEMS syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4063 C Reactive Protein from human plasma, buffered aqueous solution