EMAIL THIS PAGE TO A FRIEND
Gene
Identification and validation of reference genes for normalization of gene expression analysis using qRT-PCR in Helicoverpa armigera (Lepidoptera: Noctuidae).
PMID 25447918
Abstract
Recent studies have focused on determining functional genes and microRNAs in the pest Helicoverpa armigera (Lepidoptera: Noctuidae). Most of these studies used quantitative real-time PCR (qRT-PCR). Suitable reference genes are necessary to normalize gene expression data of qRT-PCR. However, a comprehensive study on the reference genes in H. armigera remains lacking. Twelve candidate reference genes of H. armigera were selected and evaluated for their expression stability under different biotic and abiotic conditions. The comprehensive stability ranking of candidate reference genes was recommended by RefFinder and the optimal number of reference genes was calculated by geNorm. Two target genes, thioredoxin (TRX) and Cu/Zn superoxide dismutase (SOD), were used to validate the selection of reference genes. Results showed that the most suitable candidate combinations of reference genes were as follows: 28S and RPS15 for developmental stages; RPS15 and RPL13 for larvae tissues; EF and RPL27 for adult tissues; GAPDH, RPL27, and β-TUB for nuclear polyhedrosis virus infection; RPS15 and RPL32 for insecticide treatment; RPS15 and RPL27 for temperature treatment; and RPL32, RPS15, and RPL27 for all samples. This study not only establishes an accurate method for normalizing qRT-PCR data in H. armigera but also serve as a reference for further study on gene transcription in H. armigera and other insects.