EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Advanced glycation end products are mitogenic signals and trigger cell cycle reentry of neurons in Alzheimer's disease brain.
PMID 25448604
Abstract
Neurons that reenter the cell cycle die rather than divide, a phenomenon that is associated with neurodegeneration in Alzheimer's disease (AD). Reexpression of cell-cycle related genes in differentiated neurons in AD might be rooted in aberrant mitogenic signaling. Because microglia and astroglia proliferate in the vicinity of amyloid plaques, it is likely that plaque components or factors secreted from plaque-activated glia induce neuronal mitogenic signaling. Advanced glycation end products (AGEs), protein-bound oxidation products of sugar, might be one of those mitogenic compounds. Cyclin D1 positive neurons are colocalized with AGEs or directly surrounded by extracellular AGE deposits in AD brain. However, a direct proof of DNA replication in these cells has been missing. Here, we report by using fluorescent in situ hybridization that consistent with the expression of cell cycle proteins, hyperploid neuronal cells are in colocalization with AGE staining in AD brains but not in nondemented controls. To complement human data, we used apolipoprotein E-deficient mice as model of neurodegeneration and showed that increased oxidative stress caused an intensified neuronal deposition of AGEs, being accompanied by an activation of the MAPK cascade via RAGE. This cascade, in turn, induced the expression of cyclin D1 and DNA replication. In addition, reduction of oxidative stress by application of α-lipoic acid decreased AGE accumulations, and this decrease was accompanied by a reduction in cell cycle reentry and a more euploid neuronal genome.