EMAIL THIS PAGE TO A FRIEND
American journal of ophthalmology
Exogenous fungal endophthalmitis: an analysis of isolates and susceptibilities to antifungal agents over a 20-year period (1990-2010).
PMID 25449001
Abstract
To describe the isolates and susceptibilities to antifungal agents for patients with culture-proven exogenous fungal endophthalmitis. Noncomparative case series. The clinical records of all patients treated for culture-proven exogenous fungal endophthalmitis at a university referral center from 1990 to 2010 were reviewed. Specimens initially used for diagnosis were recovered from the microbiology department and then underwent antifungal sensitivity analysis. The antifungal susceptibilities of 47 fungal isolates from culture-positive fungal endophthalmitis are reported. Included are 14 isolates from yeast and 33 from mold. The mean (±standard deviation) minimum inhibitory concetrations (MICs) for amphotericin B (2.6 ± 3.5 μg/mL), fluconazole (36.9 ± 30.7 μg/mL), and voriconazole (1.9 ± 2.9 μg/mL) are reported. Presumed susceptibility to oral fluconazole, intravenous amphotericin B, intravitreal amphotericin B, oral voriconazole, and intravitreal voriconazole occurred in 34.8%-43.5%, 0-8.3%, 68.8%, 69.8%, and 100% of isolates, respectively. Based on this laboratory study of isolates from exogenous fungal endophthalmitis, intravitreal voriconazole appears to provide the broadest spectrum of antifungal coverage and, as such, may be considered for empiric therapy of endophthalmitis caused by yeast or mold.