EMAIL THIS PAGE TO A FRIEND
Gastroenterology
Building bridges and providing transparency to the hepatitis C virus drug approval process.
PMID 25449025
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U1752
Uridine 5′-monophosphate, ≥98%
C9H13N2O9P