EMAIL THIS PAGE TO A FRIEND
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases
Detection of virulence genes in Escherichia coli isolated from patients with cystitis and pyelonephritis.
PMID 25449257
Abstract
Uropathogenic Escherichia coli (UPEC) is a common cause of ascending urinary tract infections including cystitis and pyelonephritis. The purpose of this study was to investigate virulence genes among Escherichia coli isolated from patients with cystitis and pyelonephritis. Between December 2012 and June 2013, 150 E. coli isolates from hospitalized patients with pyelonephritis (n = 72) and cystitis (n=78) were collected at Shahid Beheshti Hospital in Kashan. A PCR assay was used to evaluate the presence of virulence genes including pap, hly, aer, sfa, cnf, afa, traT, and pathogenicity island (PAI) markers in isolates. Of the total 150 UPEC isolates, 130 (86.7%) were found to carry the virulence genes studied. Nineteen different virulence patterns were identified. The most prevalent virulence pattern was UPEC including traT-PAI operons. The pap, traT, aer, hly, and PAI operons were more prevalent among patients with pyelonephritis than cystitis, and the sfa, afa, and cnf genes were not detected in any of the isolates. Higher virulence gene diversity was found among pyelonephritis UPEC isolates in comparison to cystitis UPEC isolates, showing that UPEC strains that cause pyelonephritis need more virulence factors.