EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Stem cell factor (SCF) protects osteoblasts from oxidative stress through activating c-Kit-Akt signaling.
PMID 25449280
Abstract
Osteoblasts regulate bone formation and remodeling, and are main target cells of oxidative stress in the progression of osteonecrosis. The stem cell factor (SCF)-c-Kit pathway plays important roles in the proliferation, differentiation and survival in a range of cell types, but little is known about its functions in osteoblasts. In this study, we found that c-Kit is functionally expressed in both osteoblastic-like MC3T3-E1 cells and primary murine osteoblasts. Its ligand SCF exerted significant cyto-protective effects against hydrogen peroxide (H₂O₂). SCF activated its receptor c-Kit in osteoblasts, which was required for its cyto-protective effects against H₂O₂. Pharmacological inhibition (by Imatinib and Dasatinib) or shRNA-mediated knockdown of c-Kit thus inhibited SCF-mediated osteoblast protection. Further investigations showed that protection by SCF against H₂O₂ was mediated via activation of c-Kit-dependent Akt pathway. Inhibition of Akt activation, through pharmacological or genetic means, suppressed SCF-mediated anti-H₂O₂ activity in osteoblasts. In summary, we have identified a new SCF-c-Kit-Akt physiologic pathway that protects osteoblasts from H₂O₂-induced damages, and might minimize the risk of osteonecrosis caused by oxidative stress.