EMAIL THIS PAGE TO A FRIEND
Journal of nutrition education and behavior
Construct validity of a single-item, self-rated question of diet quality.
PMID 25449828
Abstract
To provide evidence of the construct validity of a single-item overall diet question with a nomological network of self-rated and biometric measures of dietary intake and outcomes. The authors conducted a secondary analysis of survey and biometric data from a cross-sectional sample of urban-dwelling adults. In addition to self-rated diet quality, they examined dietary behaviors, biomarkers of intake, and related outcomes. Self-rated diet quality was treated as a continuous variable to calculate P for trend using regression analysis. Self-rated diet quality was significantly associated with variation in both subjective and objective measures of dietary intake (fruit and vegetable intake, Pxa0