EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
Sperm functional tests.
PMID 25450304
Abstract
Several semen parameters are used to discriminate the fertile male from the subfertile male. The most widely used parameters are sperm concentration, motility, progressive motility, and sperm morphology. Semen analysis is usually applied as described in the World Health Organization manual for semen analysis. In addition to a routine semen analysis, sperm functional tests have been described for many years, which in most cases are regarded as research tools and not part of the routine semen testing in an infertility clinic. In this review we report on the value of four sperm function tests: the sperm penetration assay, the sperm-zona pellucida binding tests, the acrosome reaction, and the hyaluronan binding assay. For each test we describe the current value, the indication for performing the test, how to interpret the results, and its therapeutic implications. Our data show that sperm functional assays are highly predictive of IVF outcome results and have the potential to assist in clinical decision making, especially to avoid the current long-standing treatment with IUI and to direct the patients to intracytoplasmic sperm injection without delay when sperm functional testing fails. We believe that advances in molecular biology techniques will allow us to develop simpler sperm function assays in the near future. This will undoubtedly help clinicians in optimizing male factor infertility diagnosis and treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7630
Hyaluronic acid sodium salt from bovine vitreous humor
H1876
Hyaluronic acid sodium salt from human umbilical cord
H5388
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
53747
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
49775
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
40583
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
94137
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
97616
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
42686
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
08185
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
75048
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-90,000
96144
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
75046
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-110,000
06742
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
75043
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 130,000-150,000
75044
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 150,000-300,000
00941
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
75574
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 500,000-750,000
53163
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
41897
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
80486
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,250,000-1,500,000
63357
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
02737
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,750,000-2,000,000
51967
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
73641
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
H9390
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide