EMAIL THIS PAGE TO A FRIEND
Proteomics. Clinical applications
Capillary-channeled polymer (C-CP) fibers for the rapid extraction of proteins from urine matrices prior to detection with MALDI-MS.
PMID 25450308
Abstract
While MS is a powerful tool for biomarker determinations, the high salt content and the small molecules present in urine poses incredible challenges. Separation/extraction methods must be employed for the isolation of target species at relevant concentrations. Micropipette tips packed with capillary-channeled polymer (C-CP) fibers are employed for the SPE of proteins from a synthetic and a certified urine matrix. Extractions are performed utilizing a very simple centrifugation method to spin-down species through the C-CP fiber tips. Proteins adsorb to the hydrophobic polypropylene fibers and are eluted in a solvent suitable for MALDI-MS analysis. Figures of merit are determined for representative compounds β2-microglobulin, retinol binding protein, and transferrin. The optimum protein processing included a 100 μL aqueous rinse and an elution solvent composition was 10 μL of 55:45 ACN:water (with triflouroacetic acid). MALDI-MS responses for the target proteins are improved from nondetectable levels to eventually yield LOD ranging from 5 to 180 nM in 1 μL aliquots. C-CP fiber tips offer a plethora of advantages including low materials costs, high throughput, microvolume processing, and the determination of sub-nanogram quantities of analyte; allowing determination of biomarkers that are otherwise undetectable in urine matrices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7927
Sinapic acid, ≥98%, powder
C11H12O5