EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Enhancing optical forces on fluorescent up-converting nanoparticles by surface charge tailoring.
PMID 25451550
Abstract
3D remote control of multifunctional fluorescent up-converting nanoparticles (UCNPs) using optical forces is being required for a great variety of applications including single-particle spectroscopy, single-particle intracellular sensing, controlled and selective light-activated drug delivery and light control at the nanoscale. Most of these potential applications find a serious limitation in the reduced value of optical forces (tens of fN) acting on these nanoparticles, due to their reduced dimensions (typically around 10 nm). In this work, this limitation is faced and it is demonstrated that the magnitude of optical forces acting on UCNPs can be enhanced by more than one order of magnitude by a controlled modification of the particle/medium interface. In particular, substitution of cationic species at the surface by other species with higher mobility could lead to UCNPs trapping with constants comparable to those of spherical metallic nanoparticles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
338869
Ammonium fluoride, ≥99.99% trace metals basis
H4FN
216011
Ammonium fluoride, ACS reagent, ≥98.0%
H4FN
30101
Ammonium fluoride, puriss. p.a., ACS reagent, ≥98%
H4FN
40292
Ammonium fluoride solution, semiconductor grade VLSI PURANAL (Honeywell 17600), 40%
H4FN
W302600
Sodium citrate dihydrate, ≥99%, FG
C6H5Na3O7 · 2H2O
S4641
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
C6H5Na3O7 · 2H2O
C7254
Sodium citrate tribasic dihydrate, ≥98%
C6H5Na3O7 · 2H2O
C8532
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
C6H5Na3O7 · 2H2O
C3434
Sodium citrate tribasic dihydrate, BioXtra, ≥99.0% (titration)
C6H5Na3O7 · 2H2O
71402
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology, ≥99.5% (NT)
C6H5Na3O7 · 2H2O
C0909
Sodium citrate tribasic dihydrate, suitable for amino acid analysis, ≥98% (titration), powder
C6H5Na3O7 · 2H2O
C5920
Sodium citrate tribasic dihydrate, insect cell culture tested
C6H5Na3O7 · 2H2O
71404
Sodium citrate tribasic dihydrate, BioUltra, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71405
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71406
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71409
Sodium citrate tribasic dihydrate, tested according to Ph.Eur.
C6H5Na3O7 · 2H2O
S1804
Trisodium citrate dihydrate, meets USP testing specifications
C6H5Na3O7 · 2H2O