EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
APE1 promotes antioxidant capacity by regulating Nrf-2 function through a redox-dependent mechanism.
PMID 25452143
Abstract
APE1 is a multifunctional protein that has recently been implicated in protecting cells from oxidative stress. In the current study, we confirmed that APE1׳s effect on cellular antioxidant capacity is related to its redox activity through the use of an APE1 functional mutant, and we investigated the mechanism through which this multifunctional protein affects the function of the transcription factor Nrf-2 in regulating oxidative stress-induced genes. Using a pair of mutants for both the redox activity and the acetylation-regulated activity of APE1, in vitro assays showed that the redox activity of APE1 is crucial for its nuclear association with Nrf-2 and subsequent activation of Nrf-2׳s transcription of several downstream genes during oxidative challenge. Important oxidative stress genes are affected by APE1 redox activity, including Hmox1, Gstm1, and Txnrd1. In addition, utilizing human non-small-cell lung cancer sample tissue as well as a nude mouse xenograft model, we determined that APE1 expression levels are inversely correlated to oxidative stress in vivo. These findings indicated that interference with these crucial functions of APE1 shows promise in preventing resistance to certain radiotherapies and that further research is necessary to understand APE1׳s complex roles in regulating both the basal redox status and the oxidative stress state of the cellular environment.
Related Materials