EMAIL THIS PAGE TO A FRIEND
The bone & joint journal
Adverse reaction to metal debris is more common in patients following MoM total hip replacement with a 36 mm femoral head than previously thought: results from a modern MoM follow-up programme.
PMID 25452362
Abstract
We conducted a retrospective study to assess the prevalence of adverse reactions to metal debris (ARMD) in patients operated on at our institution with metal-on-metal (MoM) total hip replacements with 36 mm heads using a Pinnacle acetabular shell. A total of 326 patients (150 males, 175 hips; 176 females, 203 hips) with a mean age of 62.7 years (28 to 85) and mean follow-up of 7.5 years (0.1 to 10.8) participating in our in-depth modern MoM follow-up programme were included in the study, which involved recording whole blood cobalt and chromium ion measurements, Oxford hip scores (OHS) and plain radiographs of the hip and targeted cross-sectional imaging. Elevated blood metal ion levels (> 5 parts per billion) were seen in 32 (16.1%) of the 199 patients who underwent unilateral replacement. At 23 months after the start of our modern MoM follow-up programme, 29 new cases of ARMD had been revealed. Hence, the nine-year survival of this cohort declined from 96% (95% CI 95 to 98) with the old surveillance routine to 86% (95% CI 82 to 90) following the new protocol. Although ARMD may not be as common in 36 mm MoM THRs as in those with larger heads, these results support the Medicines and Healthcare Products Regulatory Agency guidelines on regular reviews and further investigations, and emphasise the need for specific a follow-up programme for patients with MoM THRs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27066
Chromium, coating quality, 99.95%, grit
Cr
374849
Chromium, chips, 99.995% trace metals basis
Cr
255610
Chromium, chips, thickness ~2 mm, 99.5%
Cr
229563
Chromium, chunks, ≥99.99% trace metals basis
Cr
266264
Chromium, powder, 99.5%, −100 mesh
Cr
266299
Chromium, powder, ≥99% trace metals basis, −325 mesh
Cr
749052
Chromium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis
Cr
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF22618739
Chromium, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr