EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Immunization with the Entamoeba histolytica surface metalloprotease EhMSP-1 protects hamsters from amebic liver abscess.
PMID 25452550
Abstract
Diarrhea and amebic liver abscesses due to invasive Entamoeba histolytica infections are an important cause of morbidity and mortality in the developing world. Entamoeba histolytica adherence and cell migration, two phenotypes linked to virulence, are both aberrant in trophozoites deficient in the metallosurface protease EhMSP-1, which is a homologue of the Leishmania vaccine candidate leishmanolysin (GP63). We examined the potential of EhMSP-1 for use as a vaccine antigen to protect against amebic liver abscesses. First, existing serum samples from South Africans naturally infected with E. histolytica were examined by enzyme-linked immunosorbent assay (ELISA) for the presence of EhMSP-1-specific IgG. Nine of 12 (75%) people with anti-E. histolytica IgG also had EhMSP-1-specific IgG antibodies. We next used a hamster model of amebic liver abscess to determine the effect of immunization with a mixture of four recombinant EhMSP-1 protein fragments. EhMSP-1 immunization stimulated a robust IgG antibody response. Furthermore, EhMSP-1 immunization of hamsters reduced development of severe amebic liver abscesses following intrahepatic injection of E. histolytica by a combined rate of 68% in two independent animal experiments. Purified IgG from immunized compared to control animals bound to the surface of E. histolytica trophozoites and accelerated amebic lysis via activation of the classical complement cascade. We concluded that EhMSP-1 is a promising antigen that warrants further study to determine its full potential as a target for therapy and/or prevention of invasive amebiasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5412
o-Phenylenediamine, tablet, 20 mg substrate per tablet
C6H8N2
P9029
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
C6H8N2
78410
o-Phenylenediamine, suitable for fluorescence, ≥99.0% (NT)
C6H8N2
P23938
o-Phenylenediamine, flaked, 99.5%
C6H8N2
694975
o-Phenylenediamine, sublimed, ≥99%
C6H8N2
P6662
o-Phenylenediamine dihydrochloride, tablet, 1 mg substrate per tablet
C6H8N2 · 2HCl
P6787
o-Phenylenediamine dihydrochloride, tablet, 2 mg substrate per tablet
C6H8N2 · 2HCl
P8806
o-Phenylenediamine dihydrochloride, tablet, 3 mg substrate per tablet
C6H8N2 · 2HCl
P8787
o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet
C6H8N2 · 2HCl
P3804
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
C6H8N2 · 2HCl
P6912
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
C6H8N2 · 2HCl
P8287
o-Phenylenediamine dihydrochloride, tablet, 10 mg substrate per tablet
C6H8N2 · 2HCl
P4664
o-Phenylenediamine dihydrochloride, tablet, 15 mg substrate per tablet
C6H8N2 · 2HCl
P7288
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
C6H8N2 · 2HCl
P8412
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet
C6H8N2 · 2HCl
P1063
o-Phenylenediamine dihydrochloride, tablet, 60 mg substrate per tablet
C6H8N2 · 2HCl
P1526
o-Phenylenediamine dihydrochloride, peroxidase substrate
C6H8N2 · 2HCl