EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
18F-FDG PET and perfusion SPECT in the diagnosis of Alzheimer and Lewy body dementias.
PMID 25453043
Abstract
Brain imaging with glucose ((18)F-FDG) PET or blood flow (hexamethylpropyleneamine oxime) SPECT is widely used for the differential diagnosis of dementia, though direct comparisons to clearly establish superiority of one method have not been undertaken. Subjects with Alzheimer disease (AD; n = 38) and dementia with Lewy bodies (DLB; n = 30) and controls (n = 30) underwent (18)F-FDG PET and SPECT in balanced order. The main outcome measure was area under the curve (AUC) of receiver-operating-characteristic analysis of visual scan rating. Consensus diagnosis with (18)F-FDG PET was superior to SPECT for both dementia vs. no-dementia (AUC = 0.93 vs. 0.72, P = 0.001) and AD vs. DLB (AUC = 0.80 vs. 0.58, P = 0.005) comparisons. The sensitivity and specificity for dementia/no-dementia was 85% and 90%, respectively, for (18)F-FDG PET and 71% and 70%, respectively, for SPECT. (18)F-FDG PET was significantly superior to blood flow SPECT. We recommend (18)F-FDG PET be performed instead of perfusion SPECT for the differential diagnosis of degenerative dementia if functional imaging is indicated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5006
2-Fluoro-2-deoxy-D-glucose, glycosylation inhibitor, glucose analog
C6H11FO5